missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Manual Crimper, Steel, for Flip Top/Flip Off Seals or Flip Tear Up Seals

Crimping tools provide a reproducible, secure vial closure.

Brand:  Fisherbrand™ 20 06 0170

498.75 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11875050

  • £534.00 / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Crimper
  • Manual
  • Steel
  • Non-autoclavable
  • Adjustable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

20mm Crimper
Steel
For Flip Tear Up
20 mm
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.