Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
£62.30 - £450.00
Product Code | Brand | Description | Product Line | Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Product Line | Size | Price | Quantity & Availability | |||
10231585
|
Thermo Scientific™
C4013-100F |
Manual Crimper, Flip-Off | National | 13 mm |
£232.00
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10372525
|
Thermo Scientific™
C4020-100F |
Manual Crimper, Flip-Off | National | 20 mm |
£282.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10250535
|
Thermo Scientific™
C4028-100 |
Manual Crimper, Standard | National | 28 mm |
£307.17
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10773667
|
Thermo Scientific™
C4030-100 |
Manual Crimper, Standard | National | 30 mm |
£309.20
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10229964
|
Thermo Scientific™
C4032-100 |
Manual Crimper, Standard | National | 32 mm |
£345.00
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10259614
|
Thermo Scientific™
C4008-102 |
Manual Decrimper | National | 8 mm |
£210.04
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12852344
|
Thermo Scientific™
C4028-102 |
Manual Decrimper | National | 28 mm |
£307.17
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10301535
|
Thermo Scientific™
C4030-102 |
Manual Decrimper | National | 30 mm |
£386.24
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11528240
|
Thermo Scientific™
C4032-102 |
Manual Decrimper | National | 32 mm |
£421.00
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13488988
|
Thermo Scientific™
CR-20C |
Manual Crimper, Standard | Chromacol | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10147664
|
Thermo Scientific™
C4008-100 |
Manual Crimper, Standard | National | 8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10429454
|
Thermo Scientific™
C4012-100 |
Manual Crimper, Standard | National | 11 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11528470
|
Thermo Scientific™
60180-543 |
Manual Crimper, Standard | Thermo Scientific | 11 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11538470
|
Thermo Scientific™
60180-544 |
Manual Crimper, Standard | Thermo Scientific | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15290389
|
Thermo Scientific™
DCB-8C |
Manual Decrimper | Chromacol | 8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15188784
|
Thermo Scientific™
DCB-20C |
Manual Decrimper | Chromacol | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10566765
|
Thermo Scientific™
C4013-100 |
Manual Crimper, Standard | National | 13 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10488084
|
Thermo Scientific™
C4012-102 |
Manual Decrimper | National | 11 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10240535
|
Thermo Scientific™
C4013-102 |
Manual Decrimper | National | 13 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10279364
|
Thermo Scientific™
C4020-102 |
Manual Decrimper | National | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15280389
|
Thermo Scientific™
CR-8C |
Manual Crimper, Standard | Chromacol | 8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12400268
|
Thermo Scientific™
C4020-100 |
Manual Crimper, Standard | National | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
14860742
|
Thermo Scientific™
CR-11C |
Manual Crimper, Standard | Chromacol | 11 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11564815
|
Thermo Scientific™
60180-557 |
Manual Decrimper | Thermo Scientific | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10530303
|
Thermo Scientific™
DCR-11 |
Decapping Pliers | Chromacol | 11 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10143362
|
Thermo Scientific™
DCR-20 |
Decapping Pliers | Chromacol | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12054907
|
Thermo Scientific™
C4012-101 |
Decapping Pliers | National | 11 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10299454
|
Thermo Scientific™
C4013-101 |
Decapping Pliers | National | 13 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10066164
|
Thermo Scientific™
C4020-101 |
Decapping Pliers | National | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10200205
|
Thermo Scientific™
C4030-101 |
Decapping Pliers | National | 30 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Manual CrimpersLightweight, precision crimpers attach seals to chromatography or serum vials. Available for standard aluminum or flip-off seals.
- Factory set to properly crimp standard-thickness seals
- Adjustable handle setting screw allows proper crimps with septa of varying thickness
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
- Simply grip pliers around top of seal, squeeze and pry upward to remove
Place decrimping head squarely on seal and squeeze.
- Minimizes damage to vial rim with chromatography or serum vials
- Removes standard aluminum seals and many steel seals
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.