Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Use Thermo Scientific™ Manual Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively.
Brand:  Thermo Scientific™ 60180-CR20
323.76 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17334163
  • £382.00 / Each
Explore more special offers
Description
Description
Use Thermo Scientific™ Vial Crimpers and Decrimpers to manually attach or remove crimp caps from vials, respectively. These manual crimpers and decappers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to easily seal or remove crimp caps reliably.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 mm Crimp Vials
Stainless Steel/Aluminum
SureSTART Level 2
Manual Crimper