Learn More
MAP2 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-17490-100UL
Product Code. 17561556
  • £375.00 / 100µL
Explore more special offers
Description
Description
The MAP2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human MAP2. This antibody recognizes human, mouse, rat antigen. The MAP2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
MAP2
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Immunofluorescence
CoraLite 488
100 μL
4133
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MAP2 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech > 100 μL