missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP4K2 (GCK) Recombinant Human Protein
MAP4K2 (GCK) is a serine/ threonine protein kinase that activates the JNK pathway.
Brand: Invitrogen™ PR7802A
This item is not returnable.
View return policy
Description
Although the MAP4K2 (GCK) kinase is found in many tissues, its expression in lymphoid follicles is restricted to the cells of the germinal center, where it may participate in B-cell differentiation. The kinase may also play a role in the regulation of vesicle targeting or fusion.
Specifications
NP_004570.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
100 μg | |
RUO - research use only | |
MAP4K2 (GCK) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
95.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |