missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP4K2 (GCK) Recombinant Human Protein
MAP4K2 (GCK) is a serine/ threonine protein kinase that activates the JNK pathway.
£434.00 - £6900.00
Specifications
Accession Number | NP_004570.2 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 95.6kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11513637
|
Invitrogen™
PV4211 |
10μg |
£434.00
10µg |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468623
|
Invitrogen™
PR7802A |
100μg |
£1602.00
100µg |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10322303
|
Invitrogen™
PV4281 |
1mg |
£6900.00
1mg |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Although the MAP4K2 (GCK) kinase is found in many tissues, its expression in lymphoid follicles is restricted to the cells of the germinal center, where it may participate in B-cell differentiation. The kinase may also play a role in the regulation of vesicle targeting or fusion.
Specifications
NP_004570.2 | |
95.6kDa | |
RUO - research use only | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
MAP4K2 (GCK) | |
Human | |
His Tag |