Learn More
Gibco™ MarrowMAX™ Bone Marrow Medium

Fully-supplemented MarrowMAX™ Bone Marrow Medium for short-term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures.
Brand: Gibco™ 12260014
165.30 GBP valid until 2025-12-31
Use promo code "24342" to get your promotional price.
Description
Gibco MarrowMAX Bone Marrow Medium is a fully-supplemented medium developed for the short term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures.
MarrowMAX Bone Marrow Medium features:
- Superior performance
- Ready-to-use format
- Quality and performance testing
- Unique, optimized formulation
Superior performance
MarrowMAX Bone Marrow Medium outperforms other commercially available media containing Giant Cell Tumor (GCT) conditioned media, providing consistent performance with a higher mitotic index and superior chromosomal morphology.
Ready-to-use format
MarrowMAX Bone Marrow Medium is a frozen, 1X medium, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2–8°C for up to 60 days.
Quality and performance testing
Every lot of MarrowMAX Bone Marrow Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Bone marrow cells from the aspirate of a normal adult donor are cultured for 24 hours in MarrowMAX Bone Marrow Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot must pass a sterility test prior to release.
Unique, optimized formulation
MarrowMAX Bone Marrow Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), gentamicin, and L-glutamine. It also contains a novel human stromal cell conditioned medium that consists of a unique blend of hematopoietic growth factors. MarrowMAX Bone Marrow Medium does not contain GCT conditioned medium.
cGMP Manufacturing and Quality System
MarrowMAX Bone Marrow Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Order Info
Shipping Condition: Dry ice
Compliance
Manufactured at a cGMP compliant facility registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications
Frozen | |
Sterile-filtered | |
100 mL |
Bone Marrow Medium | |
Glutamine |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.