Learn More
IBA Lifesciences MART-1 peptide ELAGIGILTV (HLA-A*0201)
GREENER_CHOICE
Single peptide synthesized as presented by the MHC class I allele
Brand:  IBA Lifesciences 6-7007-901
Product Code. 15322126
Description
Description
  • MART-1 peptide ELAGIGILTV (HLA-A*0201) is a single peptide for stimulation of T cells.
  • The peptide from melanoma antigen recognized by T cells 1 (MART-1) is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
  • The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
  • Specifications
    Specifications
    Recommended: -25°C to -15°C
    1 mg
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    IBA Lifesciences MART-1 peptide ELAGIGILTV (HLA-A*0201) > Quantity: 1mg