Learn More
Eppendorf™ Mastercycler™ X50 (GB)
Eppendorf™ Mastercycler™ X50 (GB) elegantly combined speed, flexibility and PCR optimization for research applications and standardization. A new highly intuitive touch screen concept puts all of those benefits always at your fingertips.
Brand:  Eppendorf™ 6306000045
 View more versions of this product
Product Code. 15748419
  • £8840.22 / Each
Explore more special offers
Description
Description
  • There are various blocks to choose from, from the fast silver block to the 384-well block
  • Touch screen control panel
  • Innovative 2D gradient for modern PCR optimization
  • X50 with touchscreen can control up to nine other devices (connected cyclers do not have a touch screen control panel)
  • Flexlid™ concept: Use of various consumables thanks to automatic lid height adjustment
  • Small footprint
  • Fast PCR
  • PCR optimization
  • Standard PCR
  • Cycle Sequencing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
±15°C
Aluminum Block
33 cm
43 cm
10.7 kg
4 to 99°C
110 to 230 V
27.5 cm
X50t