missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Diaphragm Metering Pump Head

Designed for pumping through filters or into pressurized systems. Cole-Parmer™ MasterFlex™ Diaphragm Metering Pump Heads feature PTFE and glass wetted parts help maintain fluid purity.

Brand:  Masterflex™ 07090-42

Code : XX

Additional Details : Weight : 1.16000kg

 View more versions of this product

Product Code. 11877903

  • £1135.00 / Each
Estimated Shipment: 08-10-2024
to see stock.
Add to basket

Description

Description

  • Compatible with most Masterflex L/S drives
  • Ideal for pumping through filters or into pressurized systems
  • Check valve on inlet side of pump holds prime for up to two hours when pump is off
Specifications

Specifications

16.5 x 7.62 x 15.2 cm
For pumping through filters or into pressurized systems
7.62 cm
Diaphragm Metering Pump Head
PTFE
80 to 800 mL/min.
16.5 cm
1 Each
40 to 400 rpm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers