missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ I/P™ Easy-Load™ Pump Head for Precision Tubing

Accepts multiple tubing sizes for maximum flexibility. Cole-Parmer™ Masterflex™ I/P™ Easy-Load™ Pump Head for Precision Tubing offers increased tubing life and improved performance.

Brand:  Masterflex™ 77602-00

 View more versions of this product

Product Code. 15366038

  • £1330.00 / Each
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Accepts multiple tubing sizes for maximum flexibility. Masterflex™ I/P™ Easy-Load™ Pump Head for Precision Tubing offers increased tubing life and improved performance.

  • Tubing replacement and fluid changeover takes only seconds
  • Fast tubing changes minimize downtime
  • Automatic tubing retention eliminates user adjustments and makes setup easy
  • Adjustable occlusion lets you optimize pressure and suction capabilities and tubing life by adjusting the squeeze on the tubing
  • Five-screw mounting ensures pump head is mounted securely to drive
Specifications

Specifications

0.01 to 13 L/min.
3
7.9 in.
3.5 in.
7.4 in.
Stainless Steel
I/P Easy-Load Pump Heads
Polyphenylene Sulfide
1
Adjustable
201 mm
8.9 cm
187 mm
I/P 26, I/P 73, I/P 82
Cold-rolled Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.