missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
GREENER_CHOICE

Simplify sample storage with these color-coded caps and inserts, available racked in trays or in bulk to suit a variety of applications.

Brand:  Thermo Scientific™ 4470PUR

123.99 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11651990

  • £170.00 / Pack of 500
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.

Superior Cap Design

  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
  • ScrewTop caps are compatible with storage down to -196ºC

Cap Options Simplify Sample Identification

  • Cap trays feature standard microplate format compatible with automated equipment
  • Caps are available in seven color options to enable sample identification
  • Color inserts snap into cap for a secure fit; inserts are removable
  • Inserts are available in six colors

All caps and inserts are supplied free from DNA, RNase, DNase, endotoxins and cytotoxins

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Croygenic Tube Closure
Caps for ScrewTop Tube
Purple
Matrix™
Sterile
ScrewTop Tube
Not Autoclavable
Polypropylene, Silicone
Bulk, 500 Caps
Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction
500 μL and 1.0 mL tubes
Sterile & PCR Ready
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers