Learn More
Thermo Scientific™ Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Simplify sample storage with these color-coded caps and inserts, available racked in trays or in bulk to suit a variety of applications.
£19.80 - £386.00
Product Code | Brand | Color | Description | Fit for Purpose | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Description | Fit for Purpose | Price | Quantity & Availability | |||
11902261
|
Thermo Scientific™
4470BLK |
Black | Caps for ScrewTop Tube | Sterile & PCR Ready |
£177.00
Pack of 500 |
In Stock
Log in to see stock. |
|||
11932261
|
Thermo Scientific™
4470ORG |
Orange | Caps for ScrewTop Tube | Sterile & PCR Ready |
£177.00
Pack of 500 |
In Stock
Log in to see stock. |
|||
11942261
|
Thermo Scientific™
4470PIN |
Pink | Caps for ScrewTop Tube | Sterile & PCR Ready |
£165.00
Pack of 500 |
In Stock
Log in to see stock. |
|||
11865482
|
Thermo Scientific™
4477GRY |
Gray | Caps for ScrewTop Tube | Sterile & PCR Ready |
£320.00
Pack of 480 |
In Stock
Log in to see stock. |
|||
11875482
|
Thermo Scientific™
4477PIN |
Pink | Caps for ScrewTop Tube | Sterile & PCR Ready |
£320.00
Pack of 480 |
In Stock
Log in to see stock. |
|||
10736554
|
Thermo Scientific™
4470 |
Colorless | Caps for ScrewTop Tube | Sterile & PCR Ready |
£165.00
Pack of 500 |
In Stock
Log in to see stock. |
|||
11632000
|
Thermo Scientific™
4470RED |
Red | Caps for ScrewTop Tube | Sterile & PCR Ready |
£165.00
Pack of 500 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||
10780140
|
Thermo Scientific™
4470YEL |
Yellow | Caps for ScrewTop Tube | Sterile & PCR Ready |
£165.00
Pack of 500 |
In Stock
Log in to see stock. |
|||
11681970
|
Thermo Scientific™
4470GRE |
Green | Caps for ScrewTop Tube | Sterile & PCR Ready |
£170.00
Pack of 500 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||
10401908
|
Thermo Scientific™
4470WHI |
White | Caps for ScrewTop Tube | Sterile & PCR Ready |
£165.00
Pack of 500 |
In Stock
Log in to see stock. |
|||
11651990
|
Thermo Scientific™
4470PUR |
Purple | Caps for ScrewTop Tube | Sterile & PCR Ready |
£170.00
Pack of 500 |
In Stock
Log in to see stock. |
|||
11577429
|
Thermo Scientific™
4906 |
Yellow | ScrewTop Tube Cap Tray, Empty | Sterile & PCR Ready |
£75.50
Pack of 5 |
In Stock
Log in to see stock. |
|||
12865601
|
Thermo Scientific™
4477 |
Colorless | ScrewTop Tube Cap Tray | Sterile & PCR Ready |
£386.00
Pack of 480 |
In Stock
Log in to see stock. |
|||
11642000
|
Thermo Scientific™
4477RED |
Red | ScrewTop Tube Cap Tray | Sterile & PCR Ready |
£329.00
Pack of 480 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||
11612010
|
Thermo Scientific™
4477YEL |
Yellow | ScrewTop Tube Cap Tray | Sterile & PCR Ready |
£329.00
Pack of 480 |
In Stock
Log in to see stock. |
|||
10290740
|
Thermo Scientific™
4477BLU |
Blue | ScrewTop Tube Cap Tray | Sterile & PCR Ready |
£329.00
Pack of 480 |
In Stock
Log in to see stock. |
|||
11317201
|
Thermo Scientific™
4477GRE |
Green | ScrewTop Tube Cap Tray | Sterile & PCR Ready |
£329.00
Pack of 480 |
In Stock
Log in to see stock. |
|||
11895482
|
Thermo Scientific™
4477WHI |
White | ScrewTop Tube Cap Tray | Sterile & PCR Ready |
£320.00
Pack of 480 |
In Stock
Log in to see stock. |
|||
11377201
|
Thermo Scientific™
4477PUR |
Purple | ScrewTop Tube Cap Tray | Sterile & PCR Ready |
£320.00
Pack of 480 |
In Stock
Log in to see stock. |
|||
11601970
|
Thermo Scientific™
4470BLU |
Blue | Caps for ScrewTop Tube | Sterile & PCR Ready |
£165.00
Pack of 500 |
In Stock
Log in to see stock. |
|||
10040273
|
Thermo Scientific™
4471 |
Red | Bulk Inserts | Sterile & PCR Ready |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.
Highest Quality
- All caps and inserts are supplied free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
Superior Cap Design
- Gasketed caps ensure critical sample protection over the storage lifetime
- Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
- Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
- ScrewTop caps are compatible with storage down to -176°C
Cap Options Simplify Sample Identification
- Cap trays feature standard microplate format compatible with automated equipment
- Caps are available in seven color options to enable sample identification
- Color inserts snap into cap for a secure fit; inserts are removable
- Inserts are available in six colors
Specifications
Sterile & PCR Ready |