Learn More
Thermo Scientific™ Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
GREENER_CHOICE
Simplify sample storage with these color-coded caps and inserts, available racked in trays or in bulk to suit a variety of applications.
Brand:  Thermo Scientific™ 4470ORG
122.12 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 11932261
  • £177.00 / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.
Superior Cap Design
  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
  • ScrewTop caps are compatible with storage down to -196ºC
Cap Options Simplify Sample Identification
  • Cap trays feature standard microplate format compatible with automated equipment
  • Caps are available in seven color options to enable sample identification
  • Color inserts snap into cap for a secure fit; inserts are removable
  • Inserts are available in six colors
All caps and inserts are supplied free from DNA, RNase, DNase, endotoxins and cytotoxins
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Caps for ScrewTop Tube
Orange
Sterile
Sterile & PCR Ready
Polypropylene
Bulk, 500 Caps
ScrewTop Tube