Learn More
Tocris Bioscience™ MaxiPost
Potassium channel modulator; exerts subtype-specific effects
Brand:  Tocris Bioscience™ 4949/10
174.51 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18715678
  • £225.00 / 10mg
Explore more special offers
Description
Description
MaxiPost is a potassium channel modulator. Acts as a positive modulator at neuronal Kv7 channels and calcium-activated K+ channels (BKCa) in HEK293 cells. Displays negative modulatory activity at Kv7.1 channels (Ki = 3.7 μM) and GABAA receptors. Displays anxiolytic activity in vivo.
Specifications
Specifications
>98%
MaxiPost
MaxiPost
10 mg
359.7