missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MaxiPost
Potassium channel modulator; exerts subtype-specific effects
£225.00 - £819.00
Specifications
Purity | >98% |
---|---|
Product Type | MaxiPost |
Molecular Weight (g/mol) | 359.7 |
Inhibitors | MaxiPost |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18715678
|
Tocris Bioscience™
4949/10 |
10 mg |
£225.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18718599
|
Tocris Bioscience™
4949/50 |
50 mg |
£819.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium channel modulator. Acts as a positive modulator at neuronal Kv7 channels and calcium-activated K+ channels (BKCa) in HEK293 cells. Displays negative modulatory activity at Kv7.1 channels (Ki = 3.7 μM) and GABAA receptors. Displays anxiolytic activity in vivo.Specifications
>98% | |
MaxiPost | |
359.7 | |
MaxiPost |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title