missing translation for 'onlineSavingsMsg'
Learn More

MBL™ Glass Graduated Cylinder with Polyethylene Stopper

Manufactured from borosilicate glass

Brand:  MBL CRC310

412.81 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15507780

  • £465.00 / Pack of 10
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from borosilicate glass

  • Complies with ISO 4788
  • Supplied with polyethylene stopper
  • Graduations in easy to read blue enamel
  • Manufactured from borosilicate glass
  • Robust hexagonal foot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hexagonal
Round
±2 mL
DIN 12685, ISO 4788
250 mL
Polypropylene Stopper
29/32
Stopper
2 mL
Blue
Graduated Cylinder
Borosilicate Glass
B
367 mm
Blue enamel
Polypropylene
Polyethylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.