Learn More
MBL Volumetrics™ Glass Graduated Cylinder
Manufactured from chemically resistant borosilicate glass. MBL Volumetrics™ Glass Graduated Cylinders are a reliable choice for measuring liquids and solutions in the laboratory.
Brand:  MBL CRC216
83.93 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11328064
  • £118.00 / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from chemically resistant borosilicate glass. MBL Volumetrics™ Glass Graduated Cylinders are a reliable choice for measuring liquids and solutions in the laboratory.
Features
  • Easy to read graduations in blue enamel
  • Sturdy hexagonal foot
  • Complies with ISO 4788
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hexagonal
Round
±20 mL
DIN 12680, ISO 4788, BS 604
2000 mL
505 mm
20 mL
Blue
Graduated Cylinder
Borosilicate Glass
B
Measuring Liquids and Solutions in the Laboratory
Blue enamel