missing translation for 'onlineSavingsMsg'
Learn More

mCherry Rabbit anti-Tag, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26765-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16805135

  • £130.00 / 20µL
Estimated Shipment: 22-07-2024
to see stock.

Description

Description

MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0.5 of 15 minutes, allowing it to be visualised soon after translation.
Specifications

Specifications

mCherry
Polyclonal
Unconjugated
mCherry
mCherry Fusion Protein Ag25320
20 μL
Primary
Antibody
IgG
Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Rabbit
Antigen Affinity Chromatography
RUO
-20°C
Liquid
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers