missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Rabbit anti-Tag, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | mCherry |
---|---|
Concentration | 0.41 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16805135
|
Proteintech
26765-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16895125
|
Proteintech
26765-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0.5 of 15 minutes, allowing it to be visualised soon after translation.Specifications
mCherry | |
Western Blot | |
Unconjugated | |
Rabbit | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
mCherry Fusion Protein Ag25320 | |
Primary | |
-20°C |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
mCherry | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |