Learn More
Hirschmann™ Measuring Flask with Class B Polymethylpentene
Obtain precise standard dilutions. Hirschmann™ Measuring Flask with Class B Polymethylpentene is calibrated to contain a precise volume at a specific temperature. Flasks are used for precise dilutions and prepare standard solutions
£67.40 - £93.75
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
13118069
|
Hirschmann™
7560186 |
250 mL |
£93.75
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13108069
|
Hirschmann™
7560181 |
100 mL |
£90.20
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13188059
|
Hirschmann™
7560170 |
25 mL |
£67.40
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Class B
- PMP(TPX)
- Individual ring mark adjustment
- Standard ground and PP stopper
- Clear
- DIN EN ISO 1045
Specifications
PMP | |
A |