Learn More
Gibco™ Human Fibroblast Expansion Basal Medium (formerly Medium 106)

Human Fibroblast Expansion Basal Medium (formerly Medium 106) is a sterile, liquid tissue culture medium intended for use as one component in a complete culture environment for the growth of normal human dermal fibroblasts.
Brand: Gibco™ M106500
Description
This basal culture medium contains:
- Essential and non-essential amino acids
- Vitamins
- Other organic compounds
- Trace minerals
- Inorganic salts.
Human Fibroblast Expansion Basal Medium does NOT contain:
- Antibiotics
- Antimycotics
- Hormones
- Growth factors
- Proteins
This medium is HEPES and bicarbonate buffered and is designed for use in an incubator with an atmosphere of 5% CO2 / 95% air. To support the plating and long-term proliferation of human dermal fibroblasts, Human Fibroblast Expansion Basal Medium must be supplemented with Low Serum Growth Supplement (LSGS, Cat. No. S-003-10), or Low Serum Growth Supplement Kit (LSGS Kit, Cat. No. S-003-K).

Specifications
Store at 4°C. | |
Mammalian Cell Culture | |
Medium 106 | |
Sterile-filtered | |
Cascade Biologics™ | |
Room Temperature |
Fibroblast Cells | |
Liquid | |
Sterile-filtered | |
HEPES, Phenol Red | |
500 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2855779 | Certificate of Analysis | 31/01/2025 | M106500 |
2753938 | Certificate of Analysis | 29/01/2025 | M106500 |
2419003 | Certificate of Analysis | 29/01/2025 | M106500 |
2632937 | Certificate of Analysis | 29/01/2025 | M106500 |
2753940 | Certificate of Analysis | 28/01/2025 | M106500 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.