Learn More
OHAUS™ Medium Three Prong Sleeves
OHAUS™ Medium Three Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
£9.50 - £10.35
Product Code | Brand | Color | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | Price | Quantity & Availability | |||||
15782669
|
OHAUS™
30400259 |
Red | Vinyl |
£10.35
Each |
In Stock
Log in to see stock. |
|||||
15732679
|
OHAUS™
30400264 |
Black | Vinyl |
£9.50
Each |
In Stock
Log in to see stock. |
|||||
15792669
|
OHAUS™
30400260 |
Gray | Fiberglass |
£10.35
Each |
In Stock
Log in to see stock. |
|||||
Description
- Available in red or black vinyl and fiberglass
- Fiberglass sleeves are ideal for application above 100°C
Specifications
3 | |
Prong Sleeve | |
Medium |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
OHAUS™ Medium Three Prong Sleeves