missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Medium Three Prong Sleeves
OHAUS™ Medium Three Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
Brand: OHAUS™ 30400264
8.90 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in red or black vinyl and fiberglass
- Fiberglass sleeves are ideal for application above 100°C

Specifications
3 | |
Vinyl | |
Black |
Prong Sleeve | |
Medium |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
OHAUS™ Medium Three Prong Sleeves > Marerial: Vinyl; Color: Black
Spot an opportunity for improvement?Share a Content Correction