Learn More
3M™ Melamine Floor Pads, Gray/White
Specially designed for use on stone, concrete, VCT, marble, and other porous floor surfaces. These highly effective pads make light work of ingrained dirt, and unsightly scuff marks. Environmentally preferable since no cleaning additives or chemicals required.
Brand:  3M™ 7000032549
353.50 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 17138570
  • £428.00 / Pack of 5
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Floor pad for modern micro-porous floor surfaces such as ceramic, terracotta, marble, and concrete
  • Cleans most dirt and marks with water alone, no cleaning additives or chemicals required
  • Environmentally preferable
  • Cleans up to 1000 m2 for single disc cleaning machines and 10,000 m2 for scrubber driers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Floor Pads
432 mm
Porous Floor Surfaces
Gray, White
Cleaning