Learn More
Gibco™ MEM Non-Essential Amino Acids Solution (100X)
Gibco™ MEM Non-Essential Amino Acids are used as a supplement for cell culture medium, to increase cell growth and viability.
£17.97
Specifications
Content And Storage | Store at 2–8°C. Protect from light. |
---|---|
Cell Type | Mammalian Cells |
Concentration | 100X |
Form | Liquid |
Product Type | Amino Acid Supplement |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11350912
|
Gibco™
11140050 |
100 mL |
£17.97
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gibco™ MEM Non-Essential Amino Acids are used as a supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Non-Essential Amino Acids contains the same non-essential amino acids found in the standard Minimum Essential Medium (MEM) at a strength of 100X. The complete formulation is available.
Specifications
Store at 2–8°C. Protect from light. | |
100X | |
Amino Acid Supplement | |
Sustainable packaging | |
0.9 |
Mammalian Cells | |
Liquid | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Safety and Handling
missing translation for 'shelfLife' : 18 Months
- MEM NON-ESSENTIAL AMINO ACIDS SOLUTION, (100X)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : amino acids
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.