missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Meningococcus Agglutinating Sera

Serologically identify Neisseria meningitidis by rapid agglutination test.

Brand:  Thermo Scientific™ R30167101

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13265589

  • £273.00 / 2mL
Estimated Shipment: 31-07-2024
to see stock.

Description

Description

Serologically identify Neisseria meningitidis Groups A, C, D, X, Y, Z and W135 for epidemiological and diagnostic purposes with Thermo Scientific™ Remel™ Meningococcus Agglutinating Sera.

Use Meningococcus Agglutinating Sera for presumptive identification of Neisseria meningitidis grown on culture media.

  • Simple, easy-to-use tests
  • Identifies major groups of Neisseria meningitidis serotypes
  • Provides accurate, reliable and rapid results
  • Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate with bacteria carrying homologous antigens.

    Serological tests are intended for screening purposes and should augment, not replace, culture procedures.

    The monovalent meningococcus agglutinating sera are suitable for use in slide agglutination tests. An alternative method of typing, counter immunoelectroophoresis is also described in literature1,2,3,4. The polyvalent agglutinating sera are for slide use only.

    Not all products are available for sale in all territories. Please inquire.

    Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

    Specifications

    Specifications

    2°C to 8°C
    N. gonorrhoeae
    Agglutinating Sera
    Group X
    Meningococcus Agglutinating Sera
    Serum
    Meningococcus Agglutinating Sera 1 dropper bottle
    2 mL
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers