Learn More
Thermo Scientific™ Meningococcus Agglutinating Sera
Serologically identify Neisseria meningitidis by rapid agglutination test.
£262.00 - £273.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | Meningococcus Agglutinating Sera |
Detectable Analytes | N. gonorrhoeae |
Sample Type | Serum |
Type | Agglutinating Sera |
Product Code | Brand | Description | Organism Group | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Organism Group | Price | Quantity & Availability | ||||
13235589
|
Thermo Scientific™
R30166701 |
Meningococcus Agglutinating Sera | Polyvalent Groups X-Z, W135 |
£262.00
2mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13265589
|
Thermo Scientific™
R30167101 |
Meningococcus Agglutinating Sera | Group X |
£273.00
2mL |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13275589
|
Thermo Scientific™
R30167201 |
Meningococcus Agglutinating Sera | Group Y |
£273.00
2mL |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13285589
|
Thermo Scientific™
R30167301 |
Meningococcus Agglutinating Sera | Group Z |
£273.00
2mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Serologically identify Neisseria meningitidis Groups A, C, D, X, Y, Z and W135 for epidemiological and diagnostic purposes with Thermo Scientific™ Remel™ Meningococcus Agglutinating Sera.
Use Meningococcus Agglutinating Sera for presumptive identification of Neisseria meningitidis grown on culture media.
Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate with bacteria carrying homologous antigens.
Serological tests are intended for screening purposes and should augment, not replace, culture procedures.
The monovalent meningococcus agglutinating sera are suitable for use in slide agglutination tests. An alternative method of typing, counter immunoelectroophoresis is also described in literature1,2,3,4. The polyvalent agglutinating sera are for slide use only.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
2°C to 8°C | |
N. gonorrhoeae | |
Agglutinating Sera | |
2 mL |
Meningococcus Agglutinating Sera | |
Serum | |
Meningococcus Agglutinating Sera 1 dropper bottle |