Learn More
Epredia™ Menzel™ Microscope Coverslips
Brand:  Epredia™ BB02400240A153FST0
 View more versions of this product
Product Code. 15707592
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Coverslip
Borosilicate Glass
24 mm
24 x 24 mm
1000/Pk.
Square
24 mm
#1 (0.13 to 0.16 mm)
Automated equipment