missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Menzel™ Microscope Coverslips

£16.20 - £40.35

Specifications

Product Type Coverslip
Shape Square
Material Borosilicate Glass
Thickness (Metric) #1 (0.13 to 0.16 mm)
For Use With (Equipment) Automated equipment
View More Specs

Products 3
Product Code Brand Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Price Quantity & Availability  
15767572
View Documents
Epredia™
BB01800180A153FST0
18 mm 18 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11961988
View Documents Promotion Details
Epredia™
BB02000200A153MNZ0
20 mm 20 mm
£40.35
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
 
15707592
View Documents
Epredia™
BB02400240A153FST0
24 mm 24 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

Coverslip
Borosilicate Glass
Automated equipment
Square
#1 (0.13 to 0.16 mm)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.