Learn More
Mercuric acetate, ACS reagent, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 179.0°C to 182.0°C |
Quantity | 100 g |
Linear Formula | (CH3CO2)2Hg |
Merck Index | 15,594 |
Solubility Information | Solubility in water: 400g/L (20°C). Other solubilities: soluble in alcohol |
Formula Weight | 318.67 |
Grade | ACS Reagent |
Packaging | Plastic bottle |
Chemical Name or Material | Mercuric acetate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0458463 | Certificate of Analysis | 12/12/2023 | 447271000,447270250,447275000 |
A0449033 | Certificate of Analysis | 05/01/2023 | 447271000,447270250,447275000 |
A0389403 | Certificate of Analysis | 13/12/2022 | 447271000,447270250,447275000 |
A0446685 | Certificate of Analysis | 28/10/2022 | 447271000,447270250,447275000 |
A0401629 | Certificate of Analysis | 19/04/2022 | 447271000,447270250,447275000 |
Safety and Handling



- Mercuric acetate
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
- H310-Fatal in contact with skin.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H300-Fatal if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P361-Take off immediately all contaminated clothing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.