Learn More
Mercury(I) chloride, 99+%, extra pure, Thermo Scientific Chemicals
£26.90 - £265.00
Chemical Identifiers
CAS | 10112-91-1 |
---|---|
Molecular Formula | Cl2Hg2 |
Molecular Weight (g/mol) | 472.08 |
MDL Number | MFCD00011043 |
InChI Key | ZOMNIUBKTOKEHS-UHFFFAOYSA-L |
Synonym | mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803 |
PubChem CID | 24956 |
ChEBI | CHEBI:33050 |
IUPAC Name | chloromercury |
SMILES | Cl[Hg].Cl[Hg] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10338110
|
Thermo Scientific Acros
213120050 |
5 g | Plastic bottle |
£26.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10165030
|
Thermo Scientific Acros
213121000 |
100 g | Plastic bottle |
£77.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10589813
|
Thermo Scientific Acros
213125000 |
500 g | Plastic bottle |
£265.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10112-91-1 | |
472.08 | |
ZOMNIUBKTOKEHS-UHFFFAOYSA-L | |
24956 | |
chloromercury |
Cl2Hg2 | |
MFCD00011043 | |
mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803 | |
CHEBI:33050 | |
Cl[Hg].Cl[Hg] |
Specifications
400.0°C | |
White | |
99% min. (Iodimetry) | |
Hg2Cl2 | |
15, 5962 | |
Solubility in water: practically insoluble. Other solubilities: insoluble in ether and alcohol | |
Cl[Hg].Cl[Hg] | |
472.08 | |
CHEBI:33050 | |
≥99% | |
7.15g/mL |
10112-91-1 | |
Amorphous Powder | |
Cl2Hg2 | |
MFCD00011043 | |
mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803 | |
ZOMNIUBKTOKEHS-UHFFFAOYSA-L | |
chloromercury | |
24956 | |
472.08 | |
Extra Pure | |
Mercury(I) chloride, Extra Pure |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-307-5
- Mercury(I) chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only