missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(I) chloride, 99+%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 400.0°C |
Color | White |
Physical Form | Amorphous Powder |
Quantity | 500 g |
Assay Percent Range | 99% min. (Iodimetry) |
Linear Formula | Hg2Cl2 |
Merck Index | 15, 5962 |
Solubility Information | Solubility in water: practically insoluble. Other solubilities: insoluble in ether and alcohol |
Formula Weight | 472.08 |
Percent Purity | ≥99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0461417 | Certificate of Analysis | 26/03/2024 | 213120050,213125000,213121000 |
A0303972 | Certificate of Analysis | 07/09/2023 | 213120050,213125000,213121000 |
A0454058 | Certificate of Analysis | 25/08/2023 | 213120050,213125000,213121000 |
A0424973 | Certificate of Analysis | 25/08/2023 | 213120050,213125000,213121000 |
A0417820 | Certificate of Analysis | 12/12/2022 | 213120050,213125000,213121000 |
Safety and Handling


Product Identifier
- Mercury(I) chloride
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mercury(I) chloride, 99+%, extra pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction