missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) chloride, 99.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 302.0°C |
Melting Point | 277.0°C |
Color | White |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 500 g |
Assay Percent Range | 99.4% min. (Complexometry) |
Linear Formula | HgCl2 |
Fieser | 01,652; 05,427; 06,359; 09,291; 11,467; 12,304; 13,175; 15,200 |
Merck Index | 15, 5943 |
Solubility Information | Solubility in water: 7.4 g/100 mL (20°C). Other solubilities: 1g/40 mL acetic acid, 1g/200 mL benzene - 1g/3.8 mL alcohol, 1g/22 mL ether - 1g/12 mL glycerol, soluble in acetone and ethyl acetate, slightly soluble in pyridine and cs2 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0463318 | Certificate of Analysis | 14/06/2024 | 201430250,201435000,201431000,201430010 |
A0445009 | Certificate of Analysis | 09/02/2024 | 201430250,201435000,201431000,201430010 |
A0415579 | Certificate of Analysis | 17/01/2023 | 201430250,201435000,201431000,201430010 |
A0401038 | Certificate of Analysis | 15/12/2022 | 201430250,201435000,201431000,201430010 |
A0398267 | Certificate of Analysis | 14/12/2022 | 201430250,201435000,201431000,201430010 |
Safety and Handling




Product Identifier
- Mercury(II) chloride
- Danger
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H300-Fatal if swallowed.
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mercury(II) chloride, 99.5%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction