Learn More
Mercury standard solution, for AAS, solution, 1 mg/ml Hg in 10% HNO3, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 196125000
57.48 GBP valid until 2024-12-20
Use promo code "21695" to get your promotional price.
View more versions of this product
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Mercury standard solution | |
1 mg/ml Hg in 10% HNO3 | |
Plastic bottle | |
Black to Gray | |
MFCD00149736 | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
342.62 | |
342.62 | |
1.0200g/mL | |
101.0°C | |
Soluble in water |
990 to 1010μg/mL of Hg(2+) in 2 to 5% HNO3 | |
7783-34-8, 7697-37-2, 7732-18-5 | |
500 mL | |
H2HgN2O7 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
mercury(2+) hydrate dinitrate | |
3084029 | |
AAS | |
-3.0°C | |
1.02 | |
Powder, Granules or Chunks |
Safety and Handling
- Mercury atomic absorption standard solution, 1 mg/ml Hg in 10% HNO3
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only