missing translation for 'onlineSavingsMsg'
Learn More

Mercury standard solution, for AAS, solution, 1 mg/ml Hg in 10% HNO3, Thermo Scientific Chemicals

£21.11 - £60.50

Specifications

Chemical Name or Material Mercury standard solution
Concentration 990 to 1010μg/mL of Hg(2+) in 2 to 5% HNO3
Name Note 1 mg/ml Hg in 10% HNO3
CAS 7783-34-8, 7697-37-2, 7732-18-5
Packaging Plastic bottle
View More Specs

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10710901
View Documents Promotion Details
Thermo Scientific Acros
196121000
100 mL Plastic bottle
£21.11
100mL
Please to purchase this item. Need a web account? Register with us today!
10420861
View Documents Promotion Details
Thermo Scientific Acros
196125000
500 mL Plastic bottle
£60.50
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
Specifications

Specifications

Mercury standard solution
1 mg/ml Hg in 10% HNO3
Plastic bottle
H2HgN2O7
KVICROHOONHSRH-UHFFFAOYSA-N
342.62
342.62
1.0200g/mL
101.0°C
Soluble in water
990 to 1010μg/mL of Hg(2+) in 2 to 5% HNO3
7783-34-8, 7697-37-2, 7732-18-5
Black to Gray
MFCD00149736
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
3084029
AAS
-3.0°C
1.02
Powder, Granules or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

Warning

Product Identifier
  • Mercury atomic absorption standard solution, 1 mg/ml Hg in 10% HNO3

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only