missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MET [M1250T] Recombinant Human Protein

The M1250T mutant of MET is both activating and displays altered substrate specificity.

Brand:  Invitrogen™ PR7432A

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15498583

  • £2555.00 / 100µg
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

MET (cMET) is a receptor-like tyrosine kinase whose disregulation has been linked to many types of human malignancies.

Specifications

Specifications

NP_000236.2, M1250T
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
100μg
RUO - research use only
MET M1250T
Human
His Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
53.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
MET (HGFR) Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers