missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MET [M1250T] Recombinant Human Protein
The M1250T mutant of MET is both activating and displays altered substrate specificity.
£2555.00 - £12060.00
Specifications
Accession Number | NP_000236.2, M1250T |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 53.7kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15498583
|
Invitrogen™
PR7432A |
100μg |
£2555.00
100µg |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10007092
|
Invitrogen™
PV4272 |
1mg |
£12060.00
1mg |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MET (cMET) is a receptor-like tyrosine kinase whose disregulation has been linked to many types of human malignancies.
Specifications
NP_000236.2, M1250T | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
MET (HGFR) Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
53.7kDa | |
RUO - research use only | |
MET M1250T | |
Human | |
His Tag | |
Frozen |