missing translation for 'onlineSavingsMsg'
Learn More
meta-Phosphoric acid, 65% HPO3 basis, Honeywell Fluka™ Product Code.: 15697150

meta-Phosphoric acid, 65% HPO3 basis, Honeywell Fluka™

Product Code. 15697150 Shop All Honeywell Fluka Products
500 g, Poly bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
This item is not returnable. View return policy

Product Code. 15697150

Brand: Honeywell-Fluka 79615500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum, ∼65% HPO3 basis, pieces

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37267-86-0
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonateShow MoreShow Less
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Color Colorless
Melting Point 200°c
Boiling Point 600°C
Density 2g/mL
Packaging Poly bottle
Quantity 500 g
Linear Formula (HPO3)n
Physical Form Solid
UN Number UN3260
Percent Purity 65% (HPO3 basis)
Chemical Name or Material meta-Phosphoric acid
Show More Show Less
compliance-icons
Product Identifier
  • meta-Phosphoric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
meta-Phosphoric acid, 65% HPO3 basis, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.