missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methacholine chloride, 100.4%, MP Biomedicals™
£32.35 - £375.00
Chemical Identifiers
CAS | 62-51-1 |
---|---|
Molecular Formula | C8H18ClNO2 |
Molecular Weight (g/mol) | 195.687 |
MDL Number | MFCD00011817 |
InChI Key | JHPHVAVFUYTVCL-UHFFFAOYSA-M |
Synonym | methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride |
PubChem CID | 6114 |
ChEBI | CHEBI:50142 |
IUPAC Name | 2-acetyloxypropyl(trimethyl)azanium;chloride |
SMILES | CC(C[N+](C)(C)C)OC(=O)C.[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11421691
|
MP Biomedicals
0219023105 |
5 g |
£32.35
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431691
|
MP Biomedicals
0219023110 |
10 g |
£47.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11441691
|
MP Biomedicals
0219023125 |
25 g |
£113.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11451691
|
MP Biomedicals
0219023180 |
100 g |
£375.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
62-51-1 | |
195.687 | |
JHPHVAVFUYTVCL-UHFFFAOYSA-M | |
6114 | |
2-acetyloxypropyl(trimethyl)azanium;chloride |
C8H18ClNO2 | |
MFCD00011817 | |
methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride | |
CHEBI:50142 | |
CC(C[N+](C)(C)C)OC(=O)C.[Cl-] |
Specifications
62-51-1 | |
≥98% | |
MFCD00011817 | |
JHPHVAVFUYTVCL-UHFFFAOYSA-M | |
2-acetyloxypropyl(trimethyl)azanium;chloride | |
6114 | |
195.7 | |
Methacholine chloride |
White | |
C8H18ClNO2 | |
methacholine chloride, acetyl-beta-methylcholine chloride, provocholine, mecholyl chloride, amechol, mecholine chloride, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, chloride, methacholinium chloride, 2-acetoxypropyl trimethylammonium chloride | |
CC(C[N+](C)(C)C)OC(=O)C.[Cl-] | |
195.687 | |
CHEBI:50142 | |
Powder |
Safety and Handling
Recommended Storage : Store at 0°C.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Methacholine Chloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methacholine chloride
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title