missing translation for 'onlineSavingsMsg'
Learn More
Methanol, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10626652

Methanol, Extra Pure, SLR, Fisher Chemical™

Product Code. 10626652
1 L, Amber Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Container
Plastic-coated Glass Bottle
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10626652

Brand: Fisher Chemical M/3950/15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spiritShow MoreShow Less
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Boiling Point 64.7°C
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55cp at 20°C
Formula Weight 32.04g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2528149 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog NumberM/3950/15,M/3950/27,M/3950/21S,M/3950/21,M/3950/PC17,M/3950/PB17
Lot Number2526392 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog NumberM/3950/15,M/3950/FP21,M/3950/27,M/3950/17,M/3950/21S,M/3950/PC25,M/3950/21,M/3950/PC17,M/3950/PB17,M/3950/25
Lot Number2526179 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog NumberM/3950/15,M/3950/FP21,M/3950/27,M/3950/17,M/3950/21S,M/3950/PC25,M/3950/21,M/3950/PC17,M/3950/PB17,M/3950/25
Lot Number2524359 Certificate TypeCertificate of Analysis Date06/02/2025 Catalog NumberM/3950/15,M/3950/FP21,M/3950/27,M/3950/17,M/3950/21S,M/3950/PC25,M/3950/21,M/3950/PC17,M/3950/PB17,M/3950/25
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.