missing translation for 'onlineSavingsMsg'
Learn More
Methanol, puriss., Honeywell Product Code.: 15642580

Methanol, puriss., Honeywell
BIOPHARMA

Product Code. 15642580
2.5 L
Click to view available options
Quantity:
2.5 L
20 kg
25 L
160 kg
This item is not returnable. View return policy

Product Code. 15642580

Brand: Honeywell Riedel-de Haën 242292.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spiritShow MoreShow Less
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Density 0.791 to 0.793g/mL at 20°C, 0.791g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
UN Number 1230
Beilstein 1098229
Formula Weight 32.04g/mol
Percent Purity ≥99.7% (GC)
Grade puriss.
Chemical Name or Material Methanol, Meets analytical specification of Ph Eur
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberO2410 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog Number24229-2.5L
Lot NumberO1490 Certificate TypeCertificate of Analysis Date08/10/2024 Catalog Number24229-2.5L
Lot NumberO0500 Certificate TypeCertificate of Analysis Date13/05/2024 Catalog Number24229-2.5L
Lot NumberN3410 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog Number24229-2.5L
Lot NumberN2120 Certificate TypeCertificate of Analysis Date02/11/2023 Catalog Number24229-2.5L
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol, puriss., Honeywell >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.