missing translation for 'onlineSavingsMsg'
Learn More
Methanol, Puriss. p.a., ACS Reagent, Reagent ISO, Honeywell Burdick & Jackson™ Product Code.: 16603964

Methanol, Puriss. p.a., ACS Reagent, Reagent ISO, Honeywell Burdick & Jackson™

Product Code. 16603964
190 L
Click to view available options
190 L
This item is not returnable. View return policy

Product Code. 16603964

Brand: Honeywell Burdick & Jackson 65757190L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point -98°C
CAS 67-56-1
Color Colorless
CAS Max % 1
Density 0.79 g/cm3 (20°C)
Boiling Point 64°C to 65°C (1013 hPa)
Quantity 190 L
Flash Point 11°C
Molecular Formula CH4O
Linear Formula CH3OH
Beilstein 1098229
Synonym Methyl alcohol
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Grade ACS
Chemical Name or Material Methanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol, Puriss. p.a., ACS Reagent, Reagent ISO, Honeywell Burdick & Jackson™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.