missing translation for 'onlineSavingsMsg'
Learn More

Methanol, puriss., Honeywell
BIOPHARMA

£64.79 - £1251.85

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15642580
BIOPHARMA
View Documents Promotion Details
Honeywell Riedel-de Haën
24229-2.5L
2.5 L
£64.79
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15662580
View Documents
Honeywell Riedel-de Haën
24229-25L
25 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15652580
View Documents
Honeywell Riedel-de Haën
24229-20KG
20 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15632580
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
24229-160KG
160 kg
£1251.85
160kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO

Specifications

67-56-1
0.36
725°F
CH3OH
97.68mmHg (20°C), 410mmHg (50°C)
1098229
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04g/mol
puriss.
0.791 to 0.793g/mL at 20°C, 0.791g/mL at 25°C
9°C (48.2°F)
CH4O
MFCD00004595
1230
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.04
CHEBI:17790
≥99.7% (GC)
Methanol, Meets analytical specification of Ph Eur
Videos
Safety and Handling

Safety and Handling

P210-P260-P280-P301 + P310-P311

H225-H301 + H311 + H331-H370

Danger

missing translation for 'einecsNumber' : 200-659-6

missing translation for 'rtecsNumber' : PC1400000

Product Identifier
  • Methanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications