Learn More
Thermo Scientific™ MethElute™ Reagent (TMPAH)
For accurate, sensitive on-column methylation use Thermo Scientific™ MethElute™ Reagent (TMPAH).
Brand: Thermo Scientific™ TS-49300
Description
For accurate, sensitive, on-column methylation and detection by GC analysis use Thermo Scientific™ MethElute™ Reagent (TMPAH). It is ideally suited for the quantitative methylation and detection of barbiturates, sedatives, xanthine bases, phenolic alkaloids, and phenytoin, giving a single quantitative peak for each substance. When the reagent is heated with drug-containing extracts, serum, or urine the drugs containing reactive amino, hydroxyl, and carboxyl functions will be methylated at the reactive sites.
Performance CharacteristicsAccuracy comparable or better than the UV/TLC method (GC is superior to UV/TLC when phenobarbitol and phenytoin present)
High precision, with coefficient of variation of 5% or less
High sensitivity; barbiturates deteced down to 0.2 mg/dL
Specifications
GC | |
Room Temperature | |
0.2 M | |
10 mL |
Flammable, Toxic | |
Trimethylanilinium hydroxide in Methanol | |
10 mL |
Safety and Handling
- methelute
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H331-Toxic if inhaled.
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash hands thoroughly after handling.
- P262-Do not get in eyes, on skin, or on clothing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P310-Immediately call a POISON CENTER/doctor/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methyl alcohol, N,N,N-trimethylaniliniumiodide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.