missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MethElute™ Reagent (TMPAH)

For accurate, sensitive on-column methylation use Thermo Scientific™ MethElute™ Reagent (TMPAH).

Brand:  Thermo Scientific™ TS-49301


 View more versions of this product

Product Code. 11597851

  • £256.00 / Pack of 12

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

For accurate, sensitive, on-column methylation and detection by GC analysis use Thermo Scientific™ MethElute™ Reagent (TMPAH). It is ideally suited for the quantitative methylation and detection of barbiturates, sedatives, xanthine bases, phenolic alkaloids, and phenytoin, giving a single quantitative peak for each substance. When the reagent is heated with drug-containing extracts, serum, or urine the drugs containing reactive amino, hydroxyl, and carboxyl functions will be methylated at the reactive sites.

Performance Characteristics

Accuracy comparable or better than the UV/TLC method (GC is superior to UV/TLC when phenobarbitol and phenytoin present)

High precision, with coefficient of variation of 5% or less

High sensitivity; barbiturates deteced down to 0.2 mg/dL

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GC
Room Temperature
0.2 M
12 x 1 mL
Flammable, Toxic
Trimethylanilinium hydroxide in Methanol
12 x 1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MethElute Reagent (TMPAH)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : methanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.