missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2,5-difluoro-4-nitrobenzoate, 95%, Thermo Scientific Chemicals
£12.06 - £253.00
Chemical Identifiers
CAS | 924868-81-5 |
---|---|
Molecular Formula | C8H5F2NO4 |
Molecular Weight (g/mol) | 217.13 |
MDL Number | MFCD08689670 |
InChI Key | XBUVRWIIEREYFL-UHFFFAOYSA-N |
Synonym | methyl 2,5-difluoro-4-nitrobenzenecarboxylate, methyl2,5-difluoro-4-nitrobenzoate, benzoic acid, 2,5-difluoro-4-nitro-, methyl ester, methyldifluoronitrobenzenecarboxylate, methyl-2,5-difluoro-4-nitrobenzoate, 2,5-difluoro-4-methoxycarbonyl nitrobenzene, 2,5-difluoro-4-nitro-benzoic acid methyl ester |
PubChem CID | 18526058 |
IUPAC Name | methyl 2,5-difluoro-4-nitrobenzoate |
SMILES | COC(=O)C1=CC(F)=C(C=C1F)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15450189
|
Thermo Scientific Alfa Aesar
H63472.MD |
250 mg | |||||||
15430189
|
Thermo Scientific Alfa Aesar
H63472.03 |
1 g |
£52.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15440189
|
Thermo Scientific Alfa Aesar
H63472.06 |
5 g |
£253.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
924868-81-5 | |
217.13 | |
XBUVRWIIEREYFL-UHFFFAOYSA-N | |
18526058 | |
COC(=O)C1=CC(F)=C(C=C1F)[N+]([O-])=O |
C8H5F2NO4 | |
MFCD08689670 | |
methyl 2,5-difluoro-4-nitrobenzenecarboxylate, methyl2,5-difluoro-4-nitrobenzoate, benzoic acid, 2,5-difluoro-4-nitro-, methyl ester, methyldifluoronitrobenzenecarboxylate, methyl-2,5-difluoro-4-nitrobenzoate, 2,5-difluoro-4-methoxycarbonyl nitrobenzene, 2,5-difluoro-4-nitro-benzoic acid methyl ester | |
methyl 2,5-difluoro-4-nitrobenzoate |
Specifications
924868-81-5 | |
MFCD08689670 | |
XBUVRWIIEREYFL-UHFFFAOYSA-N | |
methyl 2,5-difluoro-4-nitrobenzoate | |
18526058 | |
95% | |
Methyl 2,5-difluoro-4-nitrobenzoate |
C8H5F2NO4 | |
methyl 2,5-difluoro-4-nitrobenzenecarboxylate, methyl2,5-difluoro-4-nitrobenzoate, benzoic acid, 2,5-difluoro-4-nitro-, methyl ester, methyldifluoronitrobenzenecarboxylate, methyl-2,5-difluoro-4-nitrobenzoate, 2,5-difluoro-4-methoxycarbonyl nitrobenzene, 2,5-difluoro-4-nitro-benzoic acid methyl ester | |
COC(=O)C1=CC(F)=C(C=C1F)[N+]([O-])=O | |
217.13 | |
217.13 | |
Solid |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only