missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-amino-5-fluorobenzoate, 98%, Thermo Scientific Chemicals
POA - £267.00
Chemical Identifiers
CAS | 884497-46-5 |
---|---|
Molecular Formula | C8H8FNO2 |
Molecular Weight (g/mol) | 169.155 |
MDL Number | MFCD05863683 |
InChI Key | YKEJVXMOMKWEPV-UHFFFAOYSA-N |
Synonym | methyl3-amino-5-fluorobenzoate, methyl 5-amino-3-fluorobenzoate, 5-amino-3-fluorobenzoic acid methyl ester, 3-amino-5-fluoro-benzoic acid methyl ester |
PubChem CID | 3807862 |
IUPAC Name | methyl 3-amino-5-fluorobenzoate |
SMILES | COC(=O)C1=CC(=CC(=C1)F)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15469488
|
Thermo Scientific Alfa Aesar
H63038.MD |
250 mg | |||||||
15449488
|
Thermo Scientific Alfa Aesar
H63038.03 |
1 g |
£66.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15459488
|
Thermo Scientific Alfa Aesar
H63038.06 |
5 g |
£267.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
884497-46-5 | |
169.155 | |
YKEJVXMOMKWEPV-UHFFFAOYSA-N | |
3807862 | |
COC(=O)C1=CC(=CC(=C1)F)N |
C8H8FNO2 | |
MFCD05863683 | |
methyl3-amino-5-fluorobenzoate, methyl 5-amino-3-fluorobenzoate, 5-amino-3-fluorobenzoic acid methyl ester, 3-amino-5-fluoro-benzoic acid methyl ester | |
methyl 3-amino-5-fluorobenzoate |
Specifications
884497-46-5 | |
C8H8FNO2 | |
methyl3-amino-5-fluorobenzoate, methyl 5-amino-3-fluorobenzoate, 5-amino-3-fluorobenzoic acid methyl ester, 3-amino-5-fluoro-benzoic acid methyl ester | |
YKEJVXMOMKWEPV-UHFFFAOYSA-N | |
methyl 3-amino-5-fluorobenzoate | |
3807862 | |
98% | |
Methyl 3-amino-5-fluorobenzoate |
White to Yellow | |
MFCD05863683 | |
Slightly soluble in water. | |
COC(=O)C1=CC(=CC(=C1)F)N | |
169.155 | |
169.16 | |
Solid |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only