Learn More
Methyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals
£22.40 - £50.15
Identifiants chimiques
CAS | 2935-90-2 |
---|---|
Molecular Formula | C4H8O2S |
Molecular Weight (g/mol) | 120.166 |
MDL Number | MFCD00004895 |
InChI Key | LDTLDBDUBGAEDT-UHFFFAOYSA-N |
Synonym | methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz |
PubChem CID | 18050 |
IUPAC Name | methyl 3-sulfanylpropanoate |
SMILES | COC(=O)CCS |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
11415576
|
Thermo Scientific Alfa Aesar
B20718.14 |
25 g |
£22.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11425576
|
Thermo Scientific Alfa Aesar
B20718.22 |
100 g |
£50.15
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl 3-mercaptopropionate is a click chemistry reagent used in the modification of oxazolines using thiol-ene click modification. It also has implications for self-assembled monolayers (SAMs). It is used in the colorimetric detection of mercury(II) in a high-salinity solution using gold nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-mercaptopropionate is a click chemistry reagent used in the modification of oxazolines using thiol-ene click modification. It also has implications for self-assembled monolayers (SAMs). It is used in the colorimetric detection of mercury(II) in a high-salinity solution using gold nanoparticles.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Identifiants chimiques
2935-90-2 | |
120.166 | |
LDTLDBDUBGAEDT-UHFFFAOYSA-N | |
18050 | |
COC(=O)CCS |
C4H8O2S | |
MFCD00004895 | |
methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz | |
methyl 3-sulfanylpropanoate |
Spécification
2935-90-2 | |
164°C | |
C4H8O2S | |
MFCD00004895 | |
1742952 | |
Soluble in water. | |
COC(=O)CCS | |
120.166 | |
120.17 | |
Methyl 3-mercaptopropionate |
1.085 | |
60°C (140°F) | |
1.463 | |
UN1228 | |
methyl 3-mercaptopropionate, propanoic acid, 3-mercapto-, methyl ester, 3-mercaptopropionic acid methyl ester, methyl mercaptopropionate, methyl3-mercaptopropionate, methyl 3-mercaptopropanoate, propionic acid, 3-mercapto-, methyl ester, unii-v920f0bcrz, methyl-3-mercaptopropionate, v920f0bcrz | |
LDTLDBDUBGAEDT-UHFFFAOYSA-N | |
methyl 3-sulfanylpropanoate | |
18050 | |
98% |
Sécurité et traitement
GHS H Statement
H301-H331-H226-H312-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapour.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301-H312-H315-H319-H330-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 220-912-4
missing translation for 'rtecsNumber' : UA2465980
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Catégorie de danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
Mention de danger
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H410-Very toxic to aquatic life with long lasting effects.
Conseils de prudence
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only