Learn More
Methyl (R)-(-)-β-hydroxyisobutyrate, 98%, Thermo Scientific Chemicals
£38.33 - £106.19
Chemical Identifiers
CAS | 72657-23-9 |
---|---|
Molecular Formula | C5H10O3 |
Molecular Weight (g/mol) | 118.13 |
MDL Number | MFCD00063450 |
InChI Key | ATCCIZURPPEVIZ-SCSAIBSYSA-N |
Synonym | r-methyl 3-hydroxy-2-methylpropanoate, r---3-hydroxy-2-methylpropionic acid methyl ester, methyl r---3-hydroxy-2-methylpropionate, --methyl d-beta-hydroxyisobutyrate, methyl r---3-hydroxyisobutyrate, roche ester, methyl r-3-hydroxy-2-methylpropionate, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2r, methyl 2r-3-hydroxy-2-methylpropanoate, r---3-hydroxyisobutyric acid methyl ester |
PubChem CID | 5324733 |
IUPAC Name | methyl (2R)-3-hydroxy-2-methylpropanoate |
SMILES | CC(CO)C(=O)OC |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
72657-23-9 | |
118.13 | |
ATCCIZURPPEVIZ-SCSAIBSYSA-N | |
5324733 | |
CC(CO)C(=O)OC |
C5H10O3 | |
MFCD00063450 | |
r-methyl 3-hydroxy-2-methylpropanoate, r---3-hydroxy-2-methylpropionic acid methyl ester, methyl r---3-hydroxy-2-methylpropionate, --methyl d-beta-hydroxyisobutyrate, methyl r---3-hydroxyisobutyrate, roche ester, methyl r-3-hydroxy-2-methylpropionate, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2r, methyl 2r-3-hydroxy-2-methylpropanoate, r---3-hydroxyisobutyric acid methyl ester | |
methyl (2R)-3-hydroxy-2-methylpropanoate |
Specifications
72657-23-9 | |
1.0700g/mL | |
81°C | |
97.5% min. (GC) | |
HOCH2CH(CH3)COOCH3 | |
MFCD00063450 | |
− 23.30 | |
Solubility in water: insoluble. Other solubilities: in ethanol: soluble | |
CC(CO)C(=O)OC | |
118.13 | |
118.13 | |
Liquid |
Colorless | |
60.0°C (4.0 Torr) | |
Authentic | |
C5H10O3 | |
1.4243 | |
1.07 | |
r-methyl 3-hydroxy-2-methylpropanoate, r---3-hydroxy-2-methylpropionic acid methyl ester, methyl r---3-hydroxy-2-methylpropionate, --methyl d-beta-hydroxyisobutyrate, methyl r---3-hydroxyisobutyrate, roche ester, methyl r-3-hydroxy-2-methylpropionate, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2r, methyl 2r-3-hydroxy-2-methylpropanoate, r---3-hydroxyisobutyric acid methyl ester | |
ATCCIZURPPEVIZ-SCSAIBSYSA-N | |
methyl (2R)-3-hydroxy-2-methylpropanoate | |
5324733 | |
98% | |
Methyl (R)-(-)-β-hydroxyisobutyrate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only